Product Details

SNP ID
rs139786558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93898559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGCACACATGTTCTCAGAGCA[G/T]TCATCTATATCTGAGGTAAAAAAAA
Phenotype
MIM: 176880
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PROS1 PubMed Links

Gene Details

Gene
PROS1
Gene Name
protein S (alpha)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000313.3 1166 Missense Mutation GAA,GAC E246D NP_000304.2
NM_001314077.1 1166 Missense Mutation GAA,GAC E278D NP_001301006.1

View Full Product Details