Product Details

SNP ID
rs139865348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:20161124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTTTACCTCAAGCAGATGTGG[A/G]TTTCAAGTTTACATTTCCTACAGTC
Phenotype
MIM: 602303 MIM: 609168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT2B PubMed Links

Gene Details

Gene
KAT2B
Gene Name
lysine acetyltransferase 2B
There are no transcripts associated with this gene.

Gene
SGO1
Gene Name
shugoshin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012409.3 1933 Intron NP_001012409.1
NM_001012410.4 1933 Missense Mutation NP_001012410.1
NM_001012411.3 1933 Intron NP_001012411.1
NM_001012412.4 1933 Missense Mutation NP_001012412.1
NM_001012413.3 1933 Intron NP_001012413.1
NM_001199251.2 1933 Intron NP_001186180.1
NM_001199252.2 1933 Missense Mutation NP_001186181.1
NM_001199253.2 1933 Intron NP_001186182.1
NM_001199254.2 1933 Missense Mutation NP_001186183.1
NM_001199255.2 1933 Intron NP_001186184.1
NM_001199256.2 1933 Missense Mutation NP_001186185.1
NM_001199257.2 1933 Missense Mutation NP_001186186.1
NM_138484.4 1933 Missense Mutation NP_612493.1
XM_011533373.2 1933 Missense Mutation XP_011531675.1
XM_011533375.2 1933 Missense Mutation XP_011531677.1
XM_011533376.2 1933 Missense Mutation XP_011531678.1
XM_011533377.2 1933 Missense Mutation XP_011531679.1

View Full Product Details