Product Details

SNP ID
rs139936521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150410002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACTGCGGAGCGGGACAGCGGC[C/T]TGGGCGCCACCGGAGGGTCGGTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSC22D2 PubMed Links

Gene Details

Gene
TSC22D2
Gene Name
TSC22 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303264.1 1668 Silent Mutation CTG,TTG L218L NP_001290193.1
NM_014779.3 1668 Silent Mutation CTG,TTG L218L NP_055594.1
XM_011513337.2 1668 Silent Mutation CTG,TTG L218L XP_011511639.1

View Full Product Details