Product Details

SNP ID
rs139941796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49422416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGGAATGCCTGCAGGCGAAAG[C/T]CCAGACGGGCCACCACACTTACAGC
Phenotype
MIM: 238310 MIM: 611516 MIM: 600690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMT PubMed Links

Gene Details

Gene
AMT
Gene Name
aminomethyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000481.3 263 Missense Mutation GAC,GGC D12G NP_000472.2
NM_001164710.1 263 Missense Mutation GAC,GGC D12G NP_001158182.1
NM_001164711.1 263 Missense Mutation GAC,GGC D12G NP_001158183.1
NM_001164712.1 263 Missense Mutation GAC,GGC D12G NP_001158184.1
Gene
NICN1
Gene Name
nicolin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032316.3 263 UTR 3 NP_115692.1
Gene
TCTA
Gene Name
T-cell leukemia translocation altered
There are no transcripts associated with this gene.

View Full Product Details