Product Details

SNP ID
rs140016419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49154016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATGCAAAGCCCATGTACCTGA[C/T]AGGGAGGAGGGCAAGGCAGGCCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC71 PubMed Links

Gene Details

Gene
CCDC71
Gene Name
coiled-coil domain containing 71
There are no transcripts associated with this gene.

Gene
LAMB2P1
Gene Name
laminin subunit beta 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details