Product Details

SNP ID
rs140152218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129450450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTCAGAGAACCCCTTATGTTT[A/G]ACCAAAAAACCTTTGTATGATCTCT
Phenotype
MIM: 606045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT122 PubMed Links

Gene Details

Gene
IFT122
Gene Name
intraflagellar transport 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280541.1 Intron NP_001267470.1
NM_001280545.1 Intron NP_001267474.1
NM_001280546.1 Intron NP_001267475.1
NM_018262.3 Intron NP_060732.2
NM_052985.3 Intron NP_443711.2
NM_052989.2 Intron NP_443715.1
NM_052990.2 Intron NP_443716.1
XM_005247609.1 Intron XP_005247666.1
XM_006713689.1 Intron XP_006713752.1
XM_006713691.2 Intron XP_006713754.1
XM_006713692.2 Intron XP_006713755.1
XM_006713695.2 Intron XP_006713758.1
XM_011512972.2 Intron XP_011511274.1
XM_017006817.1 Intron XP_016862306.1
XM_017006818.1 Intron XP_016862307.1
XM_017006819.1 Intron XP_016862308.1
XM_017006820.1 Intron XP_016862309.1
XM_017006821.1 Intron XP_016862310.1
XM_017006822.1 Intron XP_016862311.1
XM_017006823.1 Intron XP_016862312.1
XM_017006824.1 Intron XP_016862313.1
XM_017006825.1 Intron XP_016862314.1
XM_017006826.1 Intron XP_016862315.1
XM_017006827.1 Intron XP_016862316.1
XM_017006828.1 Intron XP_016862317.1
XM_017006829.1 Intron XP_016862318.1
XM_017006830.1 Intron XP_016862319.1
XM_017006831.1 Intron XP_016862320.1
XM_017006832.1 Intron XP_016862321.1
XM_017006833.1 Intron XP_016862322.1
XM_017006834.1 Intron XP_016862323.1
XM_017006835.1 Intron XP_016862324.1
XM_017006836.1 Intron XP_016862325.1
XM_017006837.1 Intron XP_016862326.1
XM_017006838.1 Intron XP_016862327.1

View Full Product Details