Product Details

SNP ID
rs140361763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112533340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTTTCATTGGGGGAAACCAAT[G/T]ACTATTATGAATTATATATAGACAG
Phenotype
MIM: 609606
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATG3 PubMed Links

Gene Details

Gene
ATG3
Gene Name
autophagy related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278712.1 2227 UTR 3 NP_001265641.1
NM_022488.4 2227 Intron NP_071933.2
XM_011513074.1 2227 Intron XP_011511376.1

View Full Product Details