Product Details

SNP ID
rs140505657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121988397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCACACTCCTTCCACTATGAGA[G/T]CTGTCTTTCTCCTGGGAAATAGAAG
Phenotype
MIM: 609739
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ILDR1 PubMed Links

Gene Details

Gene
ILDR1
Gene Name
immunoglobulin like domain containing receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199799.1 1556 Missense Mutation AGA,AGC R537S NP_001186728.1
NM_001199800.1 1556 Missense Mutation AGA,AGC R448S NP_001186729.1
NM_175924.3 1556 Missense Mutation AGA,AGC R493S NP_787120.1
XM_005247389.4 1556 Missense Mutation AGA,AGC R505S XP_005247446.1
XM_011512738.2 1556 Missense Mutation ATC,CTC I524L XP_011511040.1
XM_011512739.2 1556 Missense Mutation AGA,AGC R358S XP_011511041.1

View Full Product Details