Product Details

SNP ID
rs140633242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45595182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCAAGCTGACTGTGGATGG[C/T]GCTGCCAAGCCTCCTCTTGCTGCCT
Phenotype
MIM: 604543
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIMD1 PubMed Links

Gene Details

Gene
LIMD1
Gene Name
LIM domains containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014240.2 352 Silent Mutation GGC,GGT G101G NP_055055.1
XM_011534207.2 352 Silent Mutation GGC,GGT G101G XP_011532509.1

View Full Product Details