Product Details

SNP ID
rs140695353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46500782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGTAGTTAAGCCCTGGGCCTC[A/G]GGAGAGAATGTCTATTCCTACGCAT
Phenotype
MIM: 607181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTP3 PubMed Links

Gene Details

Gene
RTP3
Gene Name
receptor transporter protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031440.1 650 Silent Mutation TCA,TCG S194S NP_113628.1

View Full Product Details