Product Details

SNP ID
rs140755249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138161893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAACAAGTAAATCATCTTAA[A/G]CTGCATCGTCATCATCATCATCCAC
Phenotype
MIM: 607024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBR1 PubMed Links

Gene Details

Gene
DBR1
Gene Name
debranching RNA lariats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016216.3 1785 Missense Mutation NP_057300.2

View Full Product Details