Product Details

SNP ID
rs140791294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:94014422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCATGGACAGCACCAACAACAC[A/G]GTACTCTGCAAGGATTTCTTTTTTT
Phenotype
MIM: 608922
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL13B PubMed Links

Gene Details

Gene
ARL13B
Gene Name
ADP ribosylation factor like GTPase 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174150.1 1105 Intron NP_001167621.1
NM_001174151.1 1105 Intron NP_001167622.1
NM_001321328.1 1105 Intron NP_001308257.1
NM_144996.3 1105 Intron NP_659433.2
NM_182896.2 1105 Intron NP_878899.1
XM_006713532.3 1105 Intron XP_006713595.1
XM_011512532.2 1105 Intron XP_011510834.1
XM_011512533.2 1105 Intron XP_011510835.1
XM_011512534.2 1105 Intron XP_011510836.1
XM_011512535.2 1105 Intron XP_011510837.1
XM_017005853.1 1105 Intron XP_016861342.1
Gene
STX19
Gene Name
syntaxin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001850.2 1105 Missense Mutation CCG,CTG P283L NP_001001850.1
XM_011512842.2 1105 Missense Mutation CCG,CTG P283L XP_011511144.1

View Full Product Details