Product Details

SNP ID
rs140905569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37053545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGAACCCGTGCCAAGGCCTC[C/T]GAGTGCCCAGTTACTGAGGCAGCTG
Phenotype
MIM: 614043 MIM: 120436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRFIP2 PubMed Links

Gene Details

Gene
LRRFIP2
Gene Name
LRR binding FLII interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134369.2 2268 UTR 3 NP_001127841.1
NM_001282691.1 2268 UTR 3 NP_001269620.1
NM_006309.3 2268 UTR 3 NP_006300.1
NM_017724.2 2268 UTR 3 NP_060194.1
XM_005265539.2 2268 UTR 3 XP_005265596.1
XM_005265540.1 2268 UTR 3 XP_005265597.1
XM_005265550.1 2268 UTR 3 XP_005265607.1
XM_005265551.1 2268 UTR 3 XP_005265608.1
XM_005265553.1 2268 UTR 3 XP_005265610.1
XM_005265554.2 2268 UTR 3 XP_005265611.1
XM_005265556.1 2268 UTR 3 XP_005265613.1
XM_006713385.1 2268 UTR 3 XP_006713448.1
XM_006713386.1 2268 UTR 3 XP_006713449.1
XM_006713387.1 2268 UTR 3 XP_006713450.1
XM_006713388.1 2268 UTR 3 XP_006713451.1
XM_006713389.1 2268 UTR 3 XP_006713452.1
XM_006713390.1 2268 UTR 3 XP_006713453.1
XM_006713392.1 2268 UTR 3 XP_006713455.1
XM_006713393.1 2268 UTR 3 XP_006713456.1
XM_006713394.1 2268 UTR 3 XP_006713457.1
XM_006713395.1 2268 UTR 3 XP_006713458.1
XM_006713396.1 2268 UTR 3 XP_006713459.1
XM_006713397.1 2268 UTR 3 XP_006713460.1
XM_006713400.1 2268 UTR 3 XP_006713463.1
XM_006713404.1 2268 UTR 3 XP_006713467.1
XM_011534217.1 2268 UTR 3 XP_011532519.1
XM_011534218.1 2268 UTR 3 XP_011532520.1
XM_011534219.1 2268 UTR 3 XP_011532521.1
XM_011534221.1 2268 UTR 3 XP_011532523.1
XM_011534222.1 2268 UTR 3 XP_011532524.1
XM_011534229.1 2268 UTR 3 XP_011532531.1
XM_017007465.1 2268 UTR 3 XP_016862954.1
XM_017007466.1 2268 UTR 3 XP_016862955.1
XM_017007467.1 2268 UTR 3 XP_016862956.1
XM_017007468.1 2268 UTR 3 XP_016862957.1
XM_017007469.1 2268 UTR 3 XP_016862958.1
XM_017007470.1 2268 UTR 3 XP_016862959.1
XM_017007471.1 2268 UTR 3 XP_016862960.1
XM_017007472.1 2268 UTR 3 XP_016862961.1
XM_017007473.1 2268 UTR 3 XP_016862962.1
XM_017007474.1 2268 UTR 3 XP_016862963.1
XM_017007475.1 2268 UTR 3 XP_016862964.1
XM_017007476.1 2268 UTR 3 XP_016862965.1
XM_017007477.1 2268 UTR 3 XP_016862966.1
XM_017007478.1 2268 UTR 3 XP_016862967.1
XM_017007479.1 2268 UTR 3 XP_016862968.1
XM_017007480.1 2268 UTR 3 XP_016862969.1
XM_017007481.1 2268 UTR 3 XP_016862970.1
XM_017007482.1 2268 UTR 3 XP_016862971.1
XM_017007483.1 2268 UTR 3 XP_016862972.1
XM_017007484.1 2268 UTR 3 XP_016862973.1
Gene
MLH1
Gene Name
mutL homolog 1
There are no transcripts associated with this gene.

View Full Product Details