Product Details

SNP ID
rs140917094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37994359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAGCGCAGGGCGCTGCGGAGGC[C/G]TTCCAGCAGCGCCTACAGGACGAGC
Phenotype
MIM: 608592
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
There are no transcripts associated with this gene.

Gene
VILL
Gene Name
villin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015873.3 584 Silent Mutation GCC,GCG A78A NP_056957.3
XM_005265191.3 584 Silent Mutation GCC,GCG A78A XP_005265248.1
XM_005265192.4 584 Silent Mutation GCC,GCG A78A XP_005265249.1
XM_006713184.3 584 Silent Mutation GCC,GCG A78A XP_006713247.1
XM_011533770.2 584 Silent Mutation GCC,GCG A78A XP_011532072.1
XM_011533771.1 584 Silent Mutation GCC,GCG A78A XP_011532073.1
XM_011533773.2 584 Silent Mutation GCC,GCG A78A XP_011532075.1

View Full Product Details