Product Details

SNP ID
rs141112061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97986825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGGGATTTTCTTCTCTTTATC[A/C]GAGATGAATCGGTGCTGGGGCTGCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GABRR3 PubMed Links

Gene Details

Gene
GABRR3
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105580.2 1379 Missense Mutation CAG,CCG Q421P NP_001099050.1

View Full Product Details