Product Details

SNP ID
rs141324034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49419098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCCTGCCAGCTTCACCTCTGG[A/G]TTTTTCAGAATAGCTGTTGCCAGGT
Phenotype
MIM: 238310 MIM: 611516 MIM: 165390 MIM: 600690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMT PubMed Links

Gene Details

Gene
AMT
Gene Name
aminomethyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000481.3 846 Silent Mutation AAC,AAT N250N NP_000472.2
NM_001164710.1 846 Silent Mutation AAC,AAT N206N NP_001158182.1
NM_001164711.1 846 Silent Mutation AAC,AAT N194N NP_001158183.1
NM_001164712.1 846 Silent Mutation AAC,AAT N250N NP_001158184.1
Gene
NICN1
Gene Name
nicolin 1
There are no transcripts associated with this gene.

Gene
RHOA
Gene Name
ras homolog family member A
There are no transcripts associated with this gene.

Gene
TCTA
Gene Name
T-cell leukemia translocation altered
There are no transcripts associated with this gene.

View Full Product Details