Product Details

SNP ID
rs141522198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37873814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAAAGATAATTCACTGTGAA[A/G]TCAAGTCTAGATCAGTAGGGCATCA
Phenotype
MIM: 608592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTDSPL PubMed Links

Gene Details

Gene
CTDSPL
Gene Name
CTD small phosphatase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008392.1 Intron NP_001008393.1
NM_005808.2 Intron NP_005799.2
XM_017005519.1 Intron XP_016861008.1
XM_017005520.1 Intron XP_016861009.1

View Full Product Details