Product Details

SNP ID
rs141525853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:17158259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGAGATGATGATGATGGGAAT[G/T]AATCTAAGCTTCATGTATACAAAAT
Phenotype
MIM: 615740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D5 PubMed Links

Gene Details

Gene
TBC1D5
Gene Name
TBC1 domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134381.1 5185 UTR 3 NP_001127853.1
NM_014744.2 5185 UTR 3 NP_055559.1
XM_005265611.1 5185 UTR 3 XP_005265668.1
XM_005265612.1 5185 UTR 3 XP_005265669.1
XM_005265614.1 5185 UTR 3 XP_005265671.1
XM_005265615.1 5185 UTR 3 XP_005265672.1
XM_005265616.4 5185 UTR 3 XP_005265673.1
XM_006713430.1 5185 UTR 3 XP_006713493.1
XM_011534281.1 5185 UTR 3 XP_011532583.1
XM_011534283.2 5185 UTR 3 XP_011532585.1
XM_011534284.2 5185 UTR 3 XP_011532586.1
XM_011534286.1 5185 UTR 3 XP_011532588.1
XM_011534287.1 5185 UTR 3 XP_011532589.1
XM_017007552.1 5185 UTR 3 XP_016863041.1
XM_017007553.1 5185 UTR 3 XP_016863042.1
XM_017007554.1 5185 UTR 3 XP_016863043.1
XM_017007555.1 5185 UTR 3 XP_016863044.1
XM_017007556.1 5185 UTR 3 XP_016863045.1
XM_017007557.1 5185 UTR 3 XP_016863046.1
XM_017007558.1 5185 UTR 3 XP_016863047.1
XM_017007559.1 5185 UTR 3 XP_016863048.1
XM_017007560.1 5185 UTR 3 XP_016863049.1
XM_017007561.1 5185 UTR 3 XP_016863050.1
XM_017007562.1 5185 UTR 3 XP_016863051.1
XM_017007563.1 5185 UTR 3 XP_016863052.1
XM_017007564.1 5185 UTR 3 XP_016863053.1
XM_017007565.1 5185 UTR 3 XP_016863054.1
XM_017007566.1 5185 UTR 3 XP_016863055.1
XM_017007567.1 5185 UTR 3 XP_016863056.1
XM_017007568.1 5185 Intron XP_016863057.1

View Full Product Details