Product Details

SNP ID
rs141595547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186666093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGTATTCGTGTGCCGTGAGTC[C/T]CACTGACTGCAGTGCTGTTGAGCCG
Phenotype
MIM: 142640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRG PubMed Links

Gene Details

Gene
HRG
Gene Name
histidine rich glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000412.4 142 Missense Mutation CCC,CTC P21L NP_000403.1
XM_005247415.3 142 Missense Mutation CCC,CTC P21L XP_005247472.1

View Full Product Details