Product Details

SNP ID
rs141656827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126105912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGTGGCATTGGCCCGAGACAGCA[C/T]GGCATCCAAGTCCCTGGAGAGAAAG
Phenotype
MIM: 600249 MIM: 610803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH1L1 PubMed Links

Gene Details

Gene
ALDH1L1
Gene Name
aldehyde dehydrogenase 1 family member L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270364.1 2718 Missense Mutation NP_001257293.1
NM_001270365.1 2718 Missense Mutation NP_001257294.1
NM_012190.3 2718 Missense Mutation NP_036322.2
XM_006713481.2 2718 Missense Mutation XP_006713544.1
XM_011512355.1 2718 Missense Mutation XP_011510657.1
XM_017005613.1 2718 Missense Mutation XP_016861102.1
XM_017005614.1 2718 Missense Mutation XP_016861103.1
Gene
ALDH1L1-AS1
Gene Name
ALDH1L1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC41A3
Gene Name
solute carrier family 41 member 3
There are no transcripts associated with this gene.

View Full Product Details