Product Details

SNP ID
rs141739731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93987317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTATTTCTGATATATTAATTTGT[A/G]TATCCGTTATATAAAAAGGAATTCT
Phenotype
MIM: 608922
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL13B PubMed Links

Gene Details

Gene
ARL13B
Gene Name
ADP ribosylation factor like GTPase 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174150.1 Intron NP_001167621.1
NM_001174151.1 Intron NP_001167622.1
NM_001321328.1 Intron NP_001308257.1
NM_144996.3 Intron NP_659433.2
NM_182896.2 Intron NP_878899.1
XM_006713532.3 Intron XP_006713595.1
XM_011512532.2 Intron XP_011510834.1
XM_011512533.2 Intron XP_011510835.1
XM_011512534.2 Intron XP_011510836.1
XM_011512535.2 Intron XP_011510837.1
XM_017005853.1 Intron XP_016861342.1

View Full Product Details