Product Details

SNP ID
rs141978531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57237538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGACCTCAAAACTTTTAAAAG[A/G]ATATGAAAAACAGGTATTGTATATC
Phenotype
MIM: 604299 MIM: 601802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 400 Missense Mutation GAA,GGA E67G NP_036228.1
XM_011533583.2 400 Missense Mutation GAA,GGA E50G XP_011531885.1
Gene
HESX1
Gene Name
HESX homeobox 1
There are no transcripts associated with this gene.

View Full Product Details