Product Details

SNP ID
rs142016683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52229064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATCCCCATTTTCACCCGGGCC[A/G]CGGATGAGGCGCTTATGGCCCCGCT
Phenotype
MIM: 605474 MIM: 607433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR9 PubMed Links

Gene Details

Gene
TLR9
Gene Name
toll like receptor 9
There are no transcripts associated with this gene.

Gene
TWF2
Gene Name
twinfilin actin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007284.3 1171 Silent Mutation CGC,CGT R340R NP_009215.1

View Full Product Details