Product Details

SNP ID
rs142061881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119469884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGAGAATTACGAACCATGTTC[A/C]AGTCAAAACTGCAGCTGCTACCATG
Phenotype
MIM: 615618
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POGLUT1 PubMed Links

Gene Details

Gene
POGLUT1
Gene Name
protein O-glucosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152305.2 180 Silent Mutation TCA,TCC S50S NP_689518.1
XM_006713705.3 180 UTR 5 XP_006713768.1
XM_017006878.1 180 UTR 5 XP_016862367.1
XM_017006879.1 180 UTR 5 XP_016862368.1
XM_017006880.1 180 UTR 5 XP_016862369.1
Gene
TMEM39A
Gene Name
transmembrane protein 39A
There are no transcripts associated with this gene.

View Full Product Details