Product Details

SNP ID
rs142077651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196707824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTAACCTCGTAAAAAACTGAA[C/T]AGCTTCTCTAGCTGCCTCAGGGATA
Phenotype
MIM: 615586 MIM: 610276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP19 PubMed Links

Gene Details

Gene
CEP19
Gene Name
centrosomal protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032898.4 391 Silent Mutation CTA,CTG L77L NP_116287.2
XM_005269370.4 391 Silent Mutation CTA,CTG L77L XP_005269427.1
XM_011513246.2 391 Silent Mutation CTA,CTG L77L XP_011511548.1
Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
There are no transcripts associated with this gene.

View Full Product Details