Product Details

SNP ID
rs142220795
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:124908270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCTCCAAGATTGGTGAAGCC[G/T]GTCGACCGCAGTTTTAGATTTTGAA
Phenotype
MIM: 147561 MIM: 612181
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ITGB5 PubMed Links

Gene Details

Gene
ITGB5
Gene Name
integrin subunit beta 5
There are no transcripts associated with this gene.

Gene
MUC13
Gene Name
mucin 13, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033049.3 1455 Silent Mutation ACA,ACC T472T NP_149038.3

View Full Product Details