Product Details

SNP ID
rs142394055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186666041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGGTTTAACAAAATGAAGGCA[C/T]TCATTGCAGCACTGCTTTTGATCAC
Phenotype
MIM: 142640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRG PubMed Links

Gene Details

Gene
HRG
Gene Name
histidine rich glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000412.4 90 Missense Mutation CTC,TTC L4F NP_000403.1
XM_005247415.3 90 Missense Mutation CTC,TTC L4F XP_005247472.1

View Full Product Details