Product Details

SNP ID
rs142444241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23208756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAGCACTAGTGGAGGAAGTTC[C/T]GATGGAGATCAACGTGAAAGTGTTC
Phenotype
MIM: 602163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2E2 PubMed Links

Gene Details

Gene
UBE2E2
Gene Name
ubiquitin conjugating enzyme E2 E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152653.3 321 Silent Mutation TCC,TCT S19S NP_689866.1
XM_005265432.2 321 Silent Mutation TCC,TCT S19S XP_005265489.1
XM_005265433.4 321 Silent Mutation TCC,TCT S19S XP_005265490.1
XM_011534074.2 321 Missense Mutation CCG,CTG P95L XP_011532376.1
XM_011534075.1 321 Missense Mutation CCG,CTG P95L XP_011532377.1
XM_011534076.2 321 Missense Mutation CCG,CTG P95L XP_011532378.1
XM_017007126.1 321 Missense Mutation CCG,CTG P95L XP_016862615.1
Gene
UBE2E2-AS1
Gene Name
UBE2E2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details