Product Details

SNP ID
rs142562211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:165188025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGCCCTTTCCAGCCGAGAAGA[C/G]AAGGGTTTCTTTGAGCCTGGCATCC
Phenotype
MIM: 609679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLITRK3 PubMed Links

Gene Details

Gene
SLITRK3
Gene Name
SLIT and NTRK like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318810.1 3250 Missense Mutation CTC,GTC L936V NP_001305739.1
NM_001318811.1 3250 Missense Mutation CTC,GTC L936V NP_001305740.1
NM_014926.3 3250 Missense Mutation CTC,GTC L936V NP_055741.2

View Full Product Details