Product Details

SNP ID
rs142829420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46373089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATAAACTGCAAAAGGCTGAAG[A/T]GCATGACTGACATCTACCTGCTCAA
Phenotype
MIM: 601373
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCR5 PubMed Links

Gene Details

Gene
CCR5
Gene Name
C-C motif chemokine receptor 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000579.3 544 Missense Mutation AGC,TGC S63C NP_000570.1
NM_001100168.1 544 Missense Mutation AGC,TGC S63C NP_001093638.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details