Product Details
- SNP ID
-
rs142859898
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:165773410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGACCCACACAACTTTCTTTCTTG[A/C]TAGTGTAATCGTTAAATTGATTTTT
- Phenotype
-
MIM: 177400
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BCHE
PubMed Links
Gene Details
- Gene
- BCHE
- Gene Name
- butyrylcholinesterase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_017006967.1 |
1943 |
Missense Mutation |
AGC,ATC |
S635I |
XP_016862456.1 |
View Full Product Details