Product Details

SNP ID
rs142871609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49804170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCTCTCCGACTTCTGCCCTCT[A/C]TTCCCCTCAGTTGTGTGGTCGGGAC
Phenotype
MIM: 191325
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDHR4 PubMed Links

Gene Details

Gene
CDHR4
Gene Name
cadherin related family member 4
There are no transcripts associated with this gene.

Gene
FAM212A
Gene Name
family with sequence similarity 212 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203370.1 Intron NP_976248.1
XM_017006372.1 Intron XP_016861861.1
Gene
MIR5193
Gene Name
microRNA 5193
There are no transcripts associated with this gene.

Gene
UBA7
Gene Name
ubiquitin like modifier activating enzyme 7
There are no transcripts associated with this gene.

View Full Product Details