Product Details

SNP ID
rs142902480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58501257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGATCCTAAGCCTTTGTATCC[C/T]ATGGATACTTTTGAAGAAGTTGTGG
Phenotype
MIM: 601641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX2 PubMed Links

Gene Details

Gene
ACOX2
Gene Name
acyl-CoA oxidase 2
There are no transcripts associated with this gene.

Gene
KCTD6
Gene Name
potassium channel tetramerization domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128214.1 1788 Silent Mutation CCC,CCT P113P NP_001121686.1
NM_153331.3 1788 Silent Mutation CCC,CCT P113P NP_699162.3
XM_005264937.2 1788 Silent Mutation CCC,CCT P113P XP_005264994.1

View Full Product Details