Product Details

SNP ID
rs142935919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9734526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATGAGGAAGCCCCCGAGGAGG[C/T]CCCTGAGAATGGCAGCAACAAGGAG
Phenotype
MIM: 602410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRPF1 PubMed Links

Gene Details

Gene
BRPF1
Gene Name
bromodomain and PHD finger containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003694.1 806 Missense Mutation GCC,GTC A129V NP_001003694.1
NM_001319049.1 806 Missense Mutation GCC,GTC A129V NP_001305978.1
NM_001319050.1 806 Missense Mutation GCC,GTC A129V NP_001305979.1
NM_004634.2 806 Missense Mutation GCC,GTC A129V NP_004625.2
XM_005265449.1 806 Missense Mutation GCC,GTC A129V XP_005265506.1
XM_005265450.1 806 Missense Mutation GCC,GTC A129V XP_005265507.1
XM_005265452.1 806 Missense Mutation GCC,GTC A129V XP_005265509.1
XM_011534101.1 806 Missense Mutation GCC,GTC A129V XP_011532403.1
XM_011534102.1 806 Missense Mutation GCC,GTC A129V XP_011532404.1
Gene
CPNE9
Gene Name
copine family member 9
There are no transcripts associated with this gene.

View Full Product Details