Product Details

SNP ID
rs143158704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194687270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTACTGCGTGGCCGACGCGCG[A/G]CTGCCCAAGGTCTTCGCCTGGGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM43A PubMed Links

Gene Details

Gene
FAM43A
Gene Name
family with sequence similarity 43 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153690.4 1378 Silent Mutation CGA,CGG R148R NP_710157.2

View Full Product Details