Product Details

SNP ID
rs143224465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:87800730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGAGCAACTGTAAACAAGCCAT[A/G]TTATGTAATACTTTACAACTACCCT
Phenotype
MIM: 182134
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR1F PubMed Links

Gene Details

Gene
HTR1F
Gene Name
5-hydroxytryptamine receptor 1F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000866.4 Intron NP_000857.1
NM_001322208.1 Intron NP_001309137.1
NM_001322209.1 Intron NP_001309138.1
NM_001322210.1 Intron NP_001309139.1
XM_005264751.3 Intron XP_005264808.1
XM_011533664.2 Intron XP_011531966.1

View Full Product Details