Product Details

SNP ID
rs143263158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12734462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACAGCTTGCAGAGCTCAGGGTC[C/T]CTTGCTGTCCTCTGAGAGCAGTGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM40 PubMed Links

Gene Details

Gene
TMEM40
Gene Name
transmembrane protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284406.1 1229 UTR 3 NP_001271335.1
NM_001284407.1 1229 UTR 3 NP_001271336.1
NM_001284408.1 1229 UTR 3 NP_001271337.1
NM_018306.3 1229 UTR 3 NP_060776.2
XM_011533937.2 1229 Intron XP_011532239.1

View Full Product Details