Product Details

SNP ID
rs143448615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97768152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGTCTTGCTTGGCCTGAAGAA[C/G]AAGGAGGTTCATGTTTTGTGCCTTG
Phenotype
MIM: 608845 MIM: 600066
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL6 PubMed Links

Gene Details

Gene
ARL6
Gene Name
ADP ribosylation factor like GTPase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006713783.3 2211 Missense Mutation AAC,AAG N15K XP_006713846.1
XM_017007311.1 2211 Missense Mutation AAC,AAG N15K XP_016862800.1
XM_017007312.1 2211 Missense Mutation AAC,AAG N15K XP_016862801.1
XM_017007313.1 2211 Missense Mutation AAC,AAG N15K XP_016862802.1
Gene
EPHA6
Gene Name
EPH receptor A6
There are no transcripts associated with this gene.

View Full Product Details