Product Details

SNP ID
rs143478451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97945269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTAGTAAGTTTCAGGTCCTTG[A/G]CAGAAATAGCTGGACTATTCCAAAT
Phenotype
MIM: 612049
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBG3 PubMed Links

Gene Details

Gene
CRYBG3
Gene Name
crystallin beta-gamma domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153605.3 1423 Intron NP_705833.3
XM_005247117.4 1423 Intron XP_005247174.1
Gene
MINA
Gene Name
MYC induced nuclear antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042533.2 1423 Missense Mutation GCC,GTC A438V NP_001035998.1
NM_001261829.1 1423 Missense Mutation GCC,GTC A437V NP_001248758.1
NM_032778.5 1423 Missense Mutation GCC,GTC A437V NP_116167.3
NM_153182.3 1423 Missense Mutation GCC,GTC A438V NP_694822.2
XM_005247838.4 1423 Missense Mutation GCC,GTC A438V XP_005247895.1
XM_011513244.2 1423 Missense Mutation GCC,GTC A207V XP_011511546.1

View Full Product Details