Product Details

SNP ID
rs143728464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52259150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAATAATTTAAACAAAGAAATG[C/T]ATAGTACCAGAGAAATAACCCCCAC
Phenotype
MIM: 612102 MIM: 608979 MIM: 611059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIRLET7G PubMed Links

Gene Details

Gene
MIRLET7G
Gene Name
microRNA let-7g
There are no transcripts associated with this gene.

Gene
PPM1M
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
There are no transcripts associated with this gene.

Gene
WDR82
Gene Name
WD repeat domain 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025222.3 Intron NP_079498.2
XM_011534136.2 Intron XP_011532438.1

View Full Product Details