Product Details

SNP ID
rs143961330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48859095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCACTACCTTCCACTCACCAG[C/T]CTGGAATCGAGACTTGAGCACATCT
Phenotype
MIM: 613698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKAR2A-AS1 PubMed Links

Gene Details

Gene
PRKAR2A-AS1
Gene Name
PRKAR2A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC25A20
Gene Name
solute carrier family 25 member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000387.5 914 Missense Mutation ACT,GCT T239A NP_000378.1

View Full Product Details