Product Details

SNP ID
rs144198819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42874353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGTAGCGGAAGCGCACATCGTG[A/G]CTGGGCTGCATGGTGCCAAAACCCC
Phenotype
MIM: 602648 MIM: 602172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACKR2 PubMed Links

Gene Details

Gene
ACKR2
Gene Name
atypical chemokine receptor 2
There are no transcripts associated with this gene.

Gene
CYP8B1
Gene Name
cytochrome P450 family 8 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004391.2 1789 Silent Mutation NP_004382.2

View Full Product Details