Product Details

SNP ID
rs144330395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33150463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTGCTAACACAGTCCCAATGCT[C/G]AGACAGAGGTTATTTTATATAGTTA
Phenotype
MIM: 605497
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
There are no transcripts associated with this gene.

Gene
SUSD5
Gene Name
sushi domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015551.1 4587 UTR 3 NP_056366.1
XM_005265034.3 4587 UTR 3 XP_005265091.1
XM_017006137.1 4587 UTR 3 XP_016861626.1

View Full Product Details