Product Details
- SNP ID
-
rs144422434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:100339165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCACTCGCGCTTGTAGCTTCATCC[A/G]GGAGGCAGCAACGCAAGGAGCCAAA
- Phenotype
-
MIM: 616769
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NIT2
PubMed Links
Gene Details
- Gene
- NIT2
- Gene Name
- nitrilase family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020202.4 |
160 |
Missense Mutation |
CAG,CGG |
Q29R |
NP_064587.1 |
View Full Product Details