Product Details

SNP ID
rs144461124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42685856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCACCTGGAGGAGGTGTCCCCG[C/G]ACGTGGTGGCCCAGGTGCTGCACTA
Phenotype
MIM: 608116 MIM: 615340
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HHATL PubMed Links

Gene Details

Gene
HHATL
Gene Name
hedgehog acyltransferase-like
There are no transcripts associated with this gene.

Gene
KLHL40
Gene Name
kelch like family member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152393.3 338 Missense Mutation CAC,GAC H80D NP_689606.2

View Full Product Details