Product Details

SNP ID
rs144606649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52064450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCTCCTCCGTCCTCCACAGAAT[A/G]AGCCCAAACTCCCACCTCTTCATGG
Phenotype
MIM: 602749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP7 PubMed Links

Gene Details

Gene
DUSP7
Gene Name
dual specificity phosphatase 7
There are no transcripts associated with this gene.

Gene
LINC00696
Gene Name
long intergenic non-protein coding RNA 696
There are no transcripts associated with this gene.

View Full Product Details