Product Details

SNP ID
rs144627905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40511473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGGAACTTGTTGTTTTAGGAA[C/T]CAGTGACCTTTGAGGATGTGGCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF620 PubMed Links

Gene Details

Gene
ZNF620
Gene Name
zinc finger protein 620
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256167.1 89 Intron NP_001243096.1
NM_001256168.1 89 Intron NP_001243097.1
NM_175888.3 89 Missense Mutation CCA,TCA P10S NP_787084.1
XM_005265011.3 89 Missense Mutation CCA,TCA P10S XP_005265068.1
XM_005265012.3 89 Missense Mutation CCA,TCA P10S XP_005265069.1
XM_017006064.1 89 Missense Mutation CCA,TCA P55S XP_016861553.1
XM_017006065.1 89 Missense Mutation CCA,TCA P10S XP_016861554.1
XM_017006066.1 89 UTR 5 XP_016861555.1
XM_017006067.1 89 UTR 5 XP_016861556.1
XM_017006068.1 89 UTR 5 XP_016861557.1
XM_017006069.1 89 Intron XP_016861558.1

View Full Product Details