Product Details

SNP ID
rs144633535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40512414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCTGGGCAGCATTCCCATTCA[C/T]CACGCCTGTTCTGGTCTCCCAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF620 PubMed Links

Gene Details

Gene
ZNF620
Gene Name
zinc finger protein 620
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256167.1 313 Intron NP_001243096.1
NM_001256168.1 313 Intron NP_001243097.1
NM_175888.3 313 Missense Mutation ACC,ATC T55I NP_787084.1
XM_005265011.3 313 Missense Mutation ACC,ATC T55I XP_005265068.1
XM_005265012.3 313 Missense Mutation ACC,ATC T55I XP_005265069.1
XM_017006064.1 313 Missense Mutation ACC,ATC T100I XP_016861553.1
XM_017006065.1 313 Intron XP_016861554.1
XM_017006066.1 313 Missense Mutation ACC,ATC T16I XP_016861555.1
XM_017006067.1 313 Missense Mutation ACC,ATC T16I XP_016861556.1
XM_017006068.1 313 Intron XP_016861557.1
XM_017006069.1 313 Intron XP_016861558.1

View Full Product Details