Product Details
- SNP ID
-
rs145084334
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:3149636 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAACAAACCAGACCAGTGAAGTCA[C/G]ACAAACCCAAAATTATTAAAGTAGT
- Phenotype
-
MIM: 609262
MIM: 612907
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRBN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199542628] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRBN
- Gene Name
- cereblon
- Gene
- TRNT1
- Gene Name
- tRNA nucleotidyl transferase 1
View Full Product Details