Product Details

SNP ID
rs145148227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187368463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATGGTTGTAGATTTCTGGACT[G/T]GGGAGCAGACATTTCAAGAACTAAT
Phenotype
MIM: 609350
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RTP4 PubMed Links

Gene Details

Gene
RTP4
Gene Name
receptor transporter protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022147.2 84 Missense Mutation GGG,TGG G8W NP_071430.2

View Full Product Details